ContactUsLinkComponent
SCSI Supercompetent Cells, Agilent Technologies
SCSI Supercompetent Cells, Agilent Technologies
# de catalogue CA99900-332
SCS1 Supercompetent Cells 5 x 0,2 ml
SCSI Supercompetent Cells, Agilent Technologies
# de catalogue CA99900-332
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    SCSI supercompetent cells
  • Size:
    5×0.2 ml
  • Cat. no.:
    CA99900-332
  • Units:
    Each
  • Details:
    5×0.2 ml; 1×10⁹ transformants/µg
Spécifications
A propos de cet article
SCSI supercompetent and DH5α competent cells are classic E. coli strains engineered for competence.
  • Engineered to become competent
This collection of competent cells, including SCSI supercompetent cells and DH5α competent cells, consists of classic E. coli strains that have been engineered to become competent for use in cloning experiments.
Caution: For research use only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought