Spécifications
- Description:TG1 electroporation-competent cells
- Size:5×0.1 ml
- Cat. no.:CAST200123
- Units:Each
- Details:For use in phage display library preparation, ≥1×10¹⁰ transformations/µg
Spécifications
A propos de cet article
Phage display libraries from electroporation-competent cells with high transformation efficiency.
- Just thaw, combine with DNA, and electroporate to get started quickly
- Efficiency of ≥1×10¹⁰ transformations/µg of pUC18 DNA
- Kit size 5×0.1 ml
Electroporation-competent cells capable of producing greater than 1×10¹⁰ transformants/µg of DNA. These electroporation-ready cells need only be thawed, mixed with DNA, and electroporated. TG1 electroporation-competent cells are recommended for preparation of phage display libraries.