Order Entry
ContactUsLinkComponent
Human Recombinant MTF2 (from Wheat germ)
Human Recombinant MTF2 (from Wheat germ)
# de catalogue 10521-768
Fournisseur:  Abnova
Numéro CAS:  
Human Recombinant MTF2 (from Wheat germ)
# de catalogue 10521-768
Fournisseur:  Abnova
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Gene ID:
    22823
  • Protein/peptide type:
    Recombinant
  • Source:
    Wheat germ
  • Species:
    Human
  • Conjugation:
    Unconjugated
  • Size:
    2 μg
  • Storage conditions:
    Store at -80C. Avoid freeze-thaw cycles.
  • Protein synonyms:
    PCL2|metal response element binding transcription factor 2
  • Protein/peptide name:
    MTF2
  • Purity:
    >80% by SDS-PAGE and Coomassie blue staining
  • Cat. no.:
    10521-768
  • Supplier No.:
    H00022823-P01

Spécifications

A propos de cet article

The Recombinant Human MTF2 Protein from Novus Biologicals is derived from Wheat germ. The Recombinant Human MTF2 Protein has been validated for the following applications: Western Blot, ELISA, SDS-Page, Protein Array, Immunoaffinity Purification.

Customers Who Bought This Also Bought