VWR® Extraction Disks, C18
Supplier: VWR International
Certificates
About this item
Economical solid phase extraction (SPE) disks that provide optimal recoveries, fast flow rates, and high solids filtering.
- Alternative for liquid/liquid extraction in EPA Method 1664
- Available with and without prefilters
- Prefilters available separately
- Reduce extraction hexane quantity up to 30%
May not be suitable for samples with high quantities of fats, silts, colloidal substances or other suspended materials.
Customers Who Bought This Also Bought