Corepoint Scientific™ White Diamond Series Undercounter and Countertop Refrigerators and Freezers, Horizon Scientific
Supplier: Horizon Scientific
Certificates
About this item
These undercounter and countertop refrigerators and freezers provide premium laboratory cold storage for scientific and medical use
- Integrated temperature alarms
- Precise microprocessor temperature control
- Forced draft circulation
- Keyed door locks
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
Freestanding units require a 4-inch margin on all sides for adequate ventilation. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Select models offer auto defrost cycles, ADA compliance, and left hinged options. For lower temperature units, -30 and -40 °C models are also available.
Certifications: *Select models ENERGY STAR certified: 76271-318, 76271-320, 76271-324, 76271-326, 76271-322, 76271-328, 76271-352, 76271-330, 76271-286, 76271-288, 76271-294, 76271-296, 76271-298, 76271-300, 76271-354, 76271-356, 76271-290, 76271-292, 76271-332, 76271-334, 76271-336, 76271-338, 76271-302, 76271-304, 76271-306, 76271-308, 76271-310, 76271-312, 76271-340, and 76271-342
Ordering information: All units have a three-year parts and labor warranty. Select models come with an additional two-year compressor parts warranty. (Please refer to unit datasheets to confirm).
Specifications
- Catalog No:
- 76271-334
- 76271-294
- 76271-338
- 76271-336
- 76271-298
- 76271-342
- 76271-296
- 76271-340
- 76271-302
- 76271-346
- 76271-300
- 76271-344
- 76271-306
- 76271-304
- 76271-310
- 76271-308
- 76271-352
- 76271-312
- 76271-356
- 76271-354
- 76271-316
- 76271-314
- 76271-320
- 76271-318
- 76271-324
- 76271-322
- 76271-328
- 76271-326
- 76271-288
- 76271-332
- 76271-286
- 76271-330
- 76271-292
- 76271-290
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76271-334
- 76271-294
- 76271-338
- 76271-336
- 76271-298
- 76271-342
- 76271-296
- 76271-340
- 76271-302
- 76271-346
- 76271-300
- 76271-344
- 76271-306
- 76271-304
- 76271-310
- 76271-308
- 76271-352
- 76271-312
- 76271-356
- 76271-354
- 76271-316
- 76271-314
- 76271-320
- 76271-318
- 76271-324
- 76271-322
- 76271-328
- 76271-326
- 76271-288
- 76271-332
- 76271-286
- 76271-330
- 76271-292
- 76271-290
- Defrost:
- Manual
- Cycle
- Auto
- Auto
- Cycle
- Manual
- Cycle
- Manual
- Manual
- Cycle
- Cycle
- Manual
- Manual
- Manual
- Manual
- Manual
- Cycle
- Manual
- Cycle
- Cycle
- Auto
- Auto
- Cycle
- Cycle
- Cycle
- Cycle
- Cycle
- Cycle
- Cycle
- Manual
- Cycle
- Cycle
- Cycle
- Cycle
- Shelves/drawers:
- 2 adjustable shelves
- 2 adjustable shelves, 1 fixed shelf
- 2 adjustable shelves
- 2 adjustable shelves
- 2 adjustable shelves, 1 fixed shelf
- 3 adjustable shelves
- 2 adjustable shelves, 1 fixed shelf
- 3 adjustable shelves
- 2 fixed shelves
- 3 adjustable shelves
- 2 adjustable shelves, 1 fixed shelf
- 2 fixed with 3 removable baskets
- 2 fixed shelves
- 2 fixed shelves
- 2 fixed shelves
- 2 fixed shelves
- 3 adjustable shelves
- 2 fixed shelves
- 2 adjustable shelves, 1 fixed shelf
- 2 adjustable shelves, 1 fixed shelf
- 3 adjustable shelves
- 3 adjustable shelves
- 2 adjustable shelves
- 2 adjustable shelves
- 2 adjustable shelves
- 4 adjustable shelves (2 large, 2 small)
- 4 adjustable shelves (2 large, 2 small)
- 2 adjustable shelves
- 2 adjustable shelves, 1 fixed shelf
- 2 adjustable shelves
- 2 adjustable shelves, 1 fixed shelf
- 3 adjustable shelves
- 2 adjustable shelves, 1 fixed shelf
- 2 adjustable shelves, 1 fixed shelf
- Door type:
- 1 solid swing door, hinge left
- 1 glass swing door, hinge right
- 1 solid swing door, hinge left
- 1 solid swing door, hinge right
- 1 glass swing door, hinge right
- 1 solid swing door, hinge right
- 1 glass swing door, hinge left
- 1 solid swing door, hinge right
- 1 solid swing door, hinge right
- 1 solid swing door, hinge right
- 1 glass swing door, hinge left
- 1 solid swing door, hinge right
- 1 solid swing door, hinge right
- 1 solid swing door, hinge left
- 1 stainless steel swing door, hinge right
- 1 solid swing door, hinge left
- 1 solid swing door, hinge right
- 1 stainless steel swing door, hinge left
- 1 solid swing door, hinge left
- 1 solid swing door, hinge right
- 1 solid swing door, hinge left
- 1 solid swing door, hinge right
- 1 solid swing door, hinge left
- 1 solid swing door, hinge right
- 1 glass swing door, hinge right
- 1 solid swing door, hinge right
- 1 glass swing door, hinge right
- 1 glass swing door, hinge left
- 1 solid swing door, hinge left
- 1 solid swing door, hinge right
- 1 solid swing door, hinge right
- 1 glass swing door, hinge right
- 1 stainless steel swing door, hinge left
- 1 stainless steel swing door, hinge right
- Electrical:
- 115 V, 60 Hz, 1.8 A ¹/₈ HP
- 115 V, 60 Hz, 1.74 A, ¹/₈ HP
- 115 V, 60 Hz, 1.8 A ¹/₈ HP
- 115 V, 60 Hz, 1 A, ¹/₈ HP
- 115 V, 60 Hz, 1.74 A, ¹/₈ HP
- 115 V, 60 Hz, 2.2 A, ¼ HP
- 115 V, 60 Hz, 1.74 A, ¹/₈ HP
- 115 V, 60 Hz, 2.2 A, ¼ HP
- 115 V, 60 Hz, 1.5 A ¹/₈ HP
- 115 V, 60 Hz, 1.2 A, ¹/₉ HP
- 115 V, 60 Hz, 1.74 A, ¹/₈ HP
- 115 V, 60 Hz, 1.4 A, ¹/₈ HP
- 115 V, 60 Hz, 1.5 A ¹/₈ HP
- 115 V, 60 Hz, 1.5 A ¹/₈ HP
- 115 V, 60 Hz, 1.5 A ¹/₈ HP
- 115 V, 60 Hz, 1.5 A ¹/₈ HP
- 115 V, 60 Hz, 0.9 A, ¹/₇ HP
- 115 V, 60 Hz, 1.5 A ¹/₈ HP
- 115 V, 60 Hz, 1.74 A, ¹/₈ HP
- 115 V, 60 Hz, 1.74 A, ¹/₈ HP
- 115 V, 60 Hz, 1.8 A ¹/₈ HP
- 115 V, 60 Hz, 1.8 A ¹/₈ HP
- 115 V, 60 Hz, 0.9 A, ¹/₈ HP
- 115 V, 60 Hz, 0.9 A, ¹/₈ HP
- 115 V, 60 Hz, 0.9 A, ¹/₈ HP
- 115 V, 60 Hz, 0.9 A, ¹/₈ HP
- 115 V, 60 Hz, 0.9 A, ¹/₈ HP
- 115 V, 60 Hz, 0.9 A, ¹/₈ HP
- 115 V, 60 Hz, 1.74 A, ¹/₈ HP
- 115 V, 60 Hz, 1 A, ¹/₈ HP
- 115 V, 60 Hz, 1.74 A, ¹/₈ HP
- 115 V, 60 Hz, 1.3 A, ¹/₆ HP
- 115 V, 60 Hz, 1.74 A, ¹/₈ HP
- 115 V, 60 Hz, 1.74 A, ¹/₈ HP
- Weight:
- 48.1 kg (106 lbs.)
- 68 kg (150 lbs.)
- 48.1 kg (106 lbs.)
- 48.1 kg (106 lbs.)
- 68 kg (150 lbs.)
- 58.1 kg (128 lbs.)
- 68 kg (150 lbs.)
- 58.1 kg (128 lbs.)
- 59.9 kg (132 lbs.)
- 53.5 kg (118 lbs.)
- 68 kg (150 lbs.)
- 53.5 kg (118 lbs.)
- 60.3 kg (133 lbs.)
- 59.9 kg (132 lbs.)
- 59.4 kg (131 lbs.)
- 60.3 kg (133 lbs.)
- 39 kg (86 lbs.)
- 59.4 kg (131 lbs.)
- 62.1 kg (137 lbs.)
- 62.1 kg (137 lbs.)
- 64.9 kg (143 lbs.)
- 64.9 kg (143 lbs.)
- 41.3 kg (91 lbs.)
- 41.3 kg (91 lbs.)
- 41.3 kg (91 lbs.)
- 38.6 kg (85 lbs.)
- 44 kg (97 lbs.)
- 41.3 kg (91 lbs.)
- 62.1 kg (137 lbs.)
- 48.1 kg (106 lbs.)
- 62.1 kg (137 lbs.)
- 58.5 kg (129 lbs.)
- 61.2 kg (135 lbs.)
- 61.2 kg (135 lbs.)
- Environmental certification:
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free
- HFC Free