A propos de cet article
Produced from sera of rabbits immunized with highly pure Recombinant Human/Murine/Rat BMP-2. AntiHuman/Murine/Rat BMP-2specific antibody was purified by affinity chromatography and then biotinylated.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Secondary
Antigen: BMP-2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human/Murine/Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 10776-814
- 10776-816
- 10776-812
- Antigen symbol:
- BMP-2
- BMP-2
- BMP-2
- Antibody type:
- Secondary
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Biotin
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG