To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Secondary
- Antigen name:Artemin
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:Artemin
- Conjugation:Biotin
- ELISA:Yes
- Size:1 mg
- Western blot:Yes
- Form:Lyophilized
- Storage buffer:See Product Data Sheet for more information.
- Molecular weight:24.2 kDa
- Storage temperature:Store at –20 °C prior to reconstitution.
- Shipping temperature:Ambient
- Immunogen:Recombinant Human Artemin
- Purification:Recombinant Human Artemin
- Packaging:PP Vial
- Supplier No.:500P245BT1MG
Spécifications
A propos de cet article
Produced from sera of rabbits immunized with highly pure Recombinant Human Artemin. AntiHuman Arteminspecific antibody was purified by affinity chromatography and then biotinylated.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Secondary
Antigen: Artemin
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human