About this item
Produced from sera of rabbits immunized with highly pure Recombinant Human Galectin-1. AntiHuman Galectin-1specific antibody was purified by affinity chromatography employing an immobilized Human Galectin-1 matrix.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: Galectin-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10776-906
- 10776-902
- 10776-904
- Antigen symbol:
- Galectin-1
- Galectin-1
- Galectin-1
- Antigen name:
- Galectin-1
- Galectin-1
- Galectin-1
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- Immunogen:
- Recombinant Human Galectin-1
- Recombinant Human Galectin-1
- Recombinant Human Galectin-1
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Molecular weight:
- 14.5 kDa
- 14.5 kDa
- 14.5 kDa
- Purification:
- Recombinant Human Galectin-1
- Recombinant Human Galectin-1
- Recombinant Human Galectin-1
- Storage buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
- Ambient