To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:BMP-7
- Clonality:Monoclonal
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:BMP-7
- Conjugation:Unconjugated
- ELISA:Yes
- Size:500 µg
- Western blot:Yes
- Form:Lyophilized
- Storage buffer:See Product Data Sheet for more information.
- Molecular weight:28.8 kDa
- Storage temperature:Store at –20 °C prior to reconstitution.
- Shipping temperature:Ambient
- Immunogen:Recombinant Human BMP-7
- Purification:Recombinant Human BMP-7
- Cat. no.:10774-918
- Packaging:Polypropylene Vial
- Supplier No.:500M123500UG
Spécifications
A propos de cet article
Monoclonal AntibodyProduced in mice immunized with highly pure Recombinant Human BMP-7. AntiHuman BMP-7specific antibody was purified from cell culture by Protein G affinity chromatography.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: BMP-7
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1k
Reactivity: Human