A propos de cet article
Produced from sera of goats immunized with highly pure Recombinant Murine TNF-α. AntiMurine TNF-αspecific antibody was purified by affinity chromatography employing an immobilized Murine TNF-α matrix.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: TNF-alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Murine
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 10775-548
- 10775-550
- 10775-546
- Antigen symbol:
- TNF-alpha
- TNF-alpha
- TNF-alpha
- Antigen name:
- TNF-alpha
- TNF-alpha
- TNF-alpha
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Reactivity:
- Mouse
- Mouse
- Mouse
- Host:
- Goat
- Goat
- Goat
- Isotype:
- IgG
- IgG
- IgG