A propos de cet article
Produced from sera of goats immunized with highly pure Recombinant Human SDF-1β (CXCL12). AntiHuman SDF-1β (CXCL12)specific antibody was purified by affinity chromatography employing an immobilized Human SDF-1β matrix.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: SDF-1beta (CXCL12)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 10775-814
- 10775-812
- 10775-810
- Antigen symbol:
- CXCL12
- CXCL12
- CXCL12
- Antigen name:
- SDF-1beta
- SDF-1beta
- SDF-1beta
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Reactivity:
- Human
- Human
- Human
- Host:
- Goat
- Goat
- Goat
- Isotype:
- IgG
- IgG
- IgG