Anti-MCP-3 (CCL7) Goat Polyclonal Antibody
Fournisseur: Peprotech
Certificats
À propos de cet article
Produced from sera of goats immunized with highly pure Recombinant Human MCP-3 (CCL7). AntiHuman MCP-3 (CCL7)specific antibody was purified by affinity chromatography employing an immobilized Human MCP-3 matrix.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: MCP-3 (CCL7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Spécifications
- Catalog No:
- 10771-268
- 10771-266
- 10771-264
- Antigen symbol:
- MCP-3 (CCL7)
- MCP-3 (CCL7)
- MCP-3 (CCL7)
- Antigen name:
- MCP-3
- MCP-3
- MCP-3
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Human
- Host:
- Goat
- Goat
- Goat
- Isotype:
- IgG
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- Immunogen:
- Recombinant Human MCP-3 (CCL7)
- Recombinant Human MCP-3 (CCL7)
- Recombinant Human MCP-3 (CCL7)
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Molecular weight:
- 9.0 kDa
- 9.0 kDa
- 9.0 kDa
- Purification:
- Recombinant Human MCP-3 (CCL7)
- Recombinant Human MCP-3 (CCL7)
- Recombinant Human MCP-3 (CCL7)
- Storage buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
- Ambient
Spécifications
Frequently Bought Together