Cole-Parmer® Stuart Digital Melting Point Apparatus, Antylia Scientific
Supplier: Argos Technologies
Certificates
About this item
No more reading glass thermometers.
- Select, measure, and display the temperature digitally, ensuring accuracy and repeatability
- Easily view the melt via magnifying lens with built-in illumination
- Be more productive—test multiple samples simultaneously
- Save time by setting a plateau temperature for initial warm-up
Designed for maximum safety and ease of use. Select, measure, and display the temperature digitally, ensuring accuracy and avoiding the need for a glass thermometer. Test two samples simultaneously, viewing the melt via a magnifying lens with built-in illumination. Extend the back feet for the optimal viewing angle.
Set a plateau temperature for initial warm-up — unattended warm-up time frees up the user until the plateau temperature is reached. Heating rate continues at 2 °C after the warm-up, slowly reaching the melt temperature for greater accuracy.
Certifications: CE approved.
BodyTextAccessories: Accessory glass capillary tubes are constructed of soda glass and easily sealed in a Bunsen flame. Supplied in a durable tube holder for added protection. Ideal for your melting point apparatus with 100 mm length, 1.9 mm OD, 1.3 mm Int.Ø, and 0.3 mm wall thickness.
Delivery: Supplied with pack of 100 capillary tubes (open at one end).
Specifications
- Catalog No:
- 76269-138
- 76269-140
- 76269-142
- Memory:
- 7 results per tube