Radiation Can Set
# de catalogue 470304-032
Fournisseur: United Scientific Supplies
Spécifications
- Description:Radiation Can Set
- Cat. no.:470304-032
Spécifications
A propos de cet article
High Quality for the Classroom.
- Includes activity guide
Two identical sized screw cap metal cans (one smooth metal and other painted dull black) to demonstrate the difference in cooling rates of hot water inside the cans due to different surfaces.
The difference in the cooling rates is attributed to the thermal radiation emitted by the two cans.
Customers Who Bought This Also Bought