Quantum Levitation Outreach Program Package
# de catalogue 470306-202
Fournisseur: QUANTUM LEVITATION LTD
Spécifications
- Description:Quantum Levitation Outreach Program Package
- Cat. no.:470306-202
Spécifications
A propos de cet article
Bring Quantum Levitation and Locking to the people.
- Easy to use
- Highly durable
The ultimate outreach package for Quantum Levitation and Quantum Locking. The kit contains portable easy-to-use experiments to be taken outside the lab and allow an hand-on experience with Quantum Levitation.
Customers Who Bought This Also Bought