Quantum Ed Levitator
# de catalogue 470306-200
Fournisseur: QUANTUM LEVITATION LTD
Spécifications
- Description:Quantum Ed Levitator
- Cat. no.:470306-200
Spécifications
A propos de cet article
The Quantum Ed Levitators are intended for science education and data analysis.
- Easy to use
- Highly durable
The levitators are equipped with a resistance thermometer for an accurate temperature measurement. The levitators are provided with a 6 months full warranty. The Quantum Ed Levitators are able to carry only their own weight + liquid nitrogen.
Customers Who Bought This Also Bought