Quantum Enhanced Levitators
Fournisseur: QUANTUM LEVITATION LTD
Certificats
A propos de cet article
The Quantum Levitators are intended for science demonstrations and visual display.
- Easy to use
- Highly durable
The levitators are provided with a 6 months full no-questions-asked warranty. The Quantum Enhanced Levitators, having extra superconducting material, are able to carry light weight (<200 g) when used in conjunction with a suitable magnetic setup or track. The X strong Enhanced Levitators can carry weight of 500 g.
Customers Who Bought This Also Bought