About this item
PeproTech's Human IL-17E ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-17E in a sandwich ELISA format.
Human IL-17E ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-17E in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human IL-17E in approximately 1000 ELISA plate wells.