A propos de cet article
PeproTech's Human/Murine/Rat BMP-2 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human/murine/rat BMP-2 in a sandwich ELISA format.
- Easy-to-use
- Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human/Murine/Rat BMP-2 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human/murine/rat BMP-2 in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human/murine/rat BMP-2 in approximately 1000 ELISA plate wells.