A propos de cet article
PeproTech's Human IL-11 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant Human IL-11 in a sandwich ELISA format.
- Easy-to-use
- Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human IL-11 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant Human IL-11 in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay Human IL-11 in approximately 1000 ELISA plate wells.