Filter Adapter Set of 7, Neoprene
# de catalogue 470303-940
Fournisseur: United Scientific Supplies
Spécifications
- Description:Filter Adapter Set of 7, Neoprene
- Cat. no.:470303-940
Spécifications
A propos de cet article
High Quality for the Classroom.
- Can be used individually or nested with adjacent sizes
- Autoclavable
Set of seven neoprene adapter cones designed to fit Buchner and fritted glass funnels, porcelain filters without headpieces, and for reverse flow cleaning of filter crucibles.
Adapters can be used to support funnels, Gooch crucibles, filter tubes, and other apparatus in filtration assemblies.
These adapters are functional even if not tightly fitted around the funnel stem. Provide an excellent alternative to boring special size holes in rubber stoppers.
Customers Who Bought This Also Bought