ContactUsLinkComponent
Esmarch Plaster Knife, OR-Grade, Sklar®
Esmarch Plaster Knife, OR-Grade, Sklar®
# de catalogue 76191-920
Fournisseur:  Sklar
Esmarch Plaster Knife Straight sharp 7in
Esmarch Plaster Knife, OR-Grade, Sklar®
# de catalogue 76191-920
Fournisseur:  Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Length:
    7"
  • Description:
    Esmarch Plaster Knife
  • Material:
    Stainless Steel
  • Curvature:
    Straight
  • Cat. no.:
    76191-920
Spécifications
A propos de cet article
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
The Esmarch Plaster Knife is a slightly curved blade used for removing pieces of a dried, plaster cast. Instrumentation used for casting is typically stronger and heavier than standard instruments in order to adequately handle the heavy cast material.
Customers Who Bought This Also Bought