Spécifications
- Length:7.75"
- Description:Hopkins Plaster Knife
- Material:Stainless Steel/Plastic
- Curvature:Straight
- Cat. no.:76191-894
Spécifications
A propos de cet article
All Sklar products are industry standard.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
The Hopkins Plaster Knife has a slightly curved blade used for removing pieces of a dried, plaster cast. Instrumentation used for casting is typically stronger and heavier than standard instruments in order to adequately handle the heavy cast material. This plaster knife has a plastic handle.