VWR® Cleanroom Papers
Supplier: VWR International
Certificates
About this item
These latex-free, cleanroom papers 28 are ideal for documentation in critical environments, where very low particle and ESD generation is important.
Certifications: Class 10 certified.
Packaging: Cleanroom product is double bagged.
Customers Who Bought This Also Bought