About this item
All Sklar products are industry standard.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Wisconsin Laryngoscope Blades are most commonly used for tracheal intubation. The Wisconsin blade is straight with a slightly curved tip to facilitate placement of an endotracheal tube. It also has a wider, C-shaped flange, which makes it useful with infants and children with large tongues.