Specifications
- Description:Caspar Screwdriver
- Length:77/8"
- Material:Stainless steel
- Cat. no.:76191-480
Specifications
About this item
All Sklar products are industry standard.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
Sklar's® Caspar Screwdriver is for use with Caspar distraction screws to hold vertebral implant plates in place. It is a pentagonal shaped screwdriver that fits over part of the screw versus simply into the tip. It has an overall length of 8".