Synthware Microliter Sample Syringes, Kemtech America
Supplier: Kemtech America
Certificates
About this item
These microliter sample syringes are available in two tip types.
- Beveled and blunt tip
- High quality borosilicate glass
- Fixed needle
The anti-coring beveled tip is suitable for easy septum penetration. The blunt tip is suitable for HPLC injection and sample pipetting.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.