Stainless Steel Micro Spoon with Plastic Handle, United Scientific Supplies
# de catalogue 76263-360
Fournisseur: United Scientific Supplies
Spécifications
- Description:Stainless Steel Micro Spoon with Plastic Handle
- Cat. no.:76263-360
Spécifications
A propos de cet article
A 15.5 cm spatula with an easy grip.
- Stainless steel construction
- Plastic coated handle
Stainless steel micro spoon has an overall length of 16.5 cm (6.5"), with one end round and one end plastic-coated for easy grip.
Customers Who Bought This Also Bought