Stainless Steel Micro Spatula, United Scientific Supplies
# de catalogue 76263-356
Fournisseur: United Scientific Supplies
Spécifications
- Length:20.3 cm (8")
- Description:Stainless steel micro spatula
- Cat. no.:76263-356
Spécifications
A propos de cet article
Stainless steel micro spatula features one end flat and one end pointed.
- Stainless steel construction
- One pointed end
This micro spatula is perfect for mixing and dispensing small volume of powders or solutions.
Customers Who Bought This Also Bought