Inspection Mirror, Industrial Physics
# de catalogue 10025-270
Fournisseur: GREAT LAKES GAGES, LLC
Spécifications
- Description:Compact telescopic mirror, glass/metal
- Cat. no.:10025-270
- Length:130/455 mm (in/out)
- Supplier No.:LD3004
- Diameter:31 mm
Spécifications
A propos de cet article
Inspection mirror spots that are difficult to inspect are even more difficult to blast or coat.
- Ideal for visual inspection of hard-to-reach spots
- With balljoint and pocket clip
With a simple inspection mirror a visual inspection behind stiffeners, under- or on top of I-beams or other places is easy to perform.
Customers Who Bought This Also Bought