Plastic Stirring Rods, United Scientific Supplies
Supplier: United Scientific Supplies
Certificates
About this item
Stirring rods are made of polypropylene, with both ends rounded.
- Polypropylene
- Autoclavable
Stirring rods are a necessary component for your laboratory. These stirring rods coming a variety of lengths and diameters fir every type of solution.
Customers Who Bought This Also Bought