Samco™ Exact Volume Transfer Pipettes, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
The exact volume transfer pipettes are used for dispensing reagents of small samples.
- Polyethylene
- Non sterile
- Ideal for home and POC (Point of Care) testing
Useful in clinical laboratories. Custom design options available to meet your specifications.
Customers Who Bought This Also Bought