Support Stands with Rods, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
All-in-one support design.
- Non-corrosive
- Steel rods are coated with polypropylene
Molded polystyrene support stands are ideal for use in applications that require non-corrosive worksurfaces. Heavy, weighted base provides stability similar to metal versions. Each comes with a rod that is 30" long and 0.5" in diameter.
Customers Who Bought This Also Bought