Plastic Spot Plate, 12 Cavities, United Scientific Supplies
# de catalogue 76263-064
Fournisseur: United Scientific Supplies
Spécifications
- Description:12 Cavity Plastic Spot Plate
- Cat. no.:76263-064
Spécifications
A propos de cet article
Economical substitute for glass or porcelain plates.
- Uniform well plates
- Easy to clean
These economical white plastic plates are suitable for most applications in which glass or porcelain plates are used. However, they are not recommended for strong solvents such as acetone or xylene. Each plate measures 112×92 mm. Supplied in a pack of 12.
Customers Who Bought This Also Bought