arium® mini plus Ultrapure Laboratory Water Systems, Sartorius (Starter Packs)
The arium® mini plus is directly connected to a feed water tap. The feed water is treated by a special designed pre-treatment cartridge, which consists of a combination of activated carbon and a reverse osmosis module.
- Simple bag exchange in less than 5 minutes
- No use of cleaning agents needed
- Optimized purity, due to closed system, avoiding a permanent biofilm
- Intuitive menu navigation with high quality glass touch-display, developed through customer survey
- Small footprint (only 28 cm width)
- Volume dispense from 50 ml
- Delivers a flow rate of 1 liter per minute
The arium® mini plus provides ASTM Types 1 and 3 water quality.
Features intuitive menu navigation for optimal user comfort. Besides the unique bagtank, the systems are equipped with a high-resolution, touch-activated color display. Easy-to-understand icons guide user intuitively through the menu. The Favorites function automatically stores the volume last dispensed. This increases efficiency and prevents errors during repeated dispensing of identical volumes.
- Catalog No:
- 75994-886
- 75994-888
- Model:
- arium® mini Plus
- arium® mini Plus
- Feed water:
- Tap water
- Tap water
- Flow rate:
- 1.0 l/min L/min
- 1.0 l/min L/min
- Production flow rate:
- 8 L/h
- 8 L/h
- Max. operating pressure:
- 0.5 - 6 bar
- 0.5 - 6 bar
- Conductivity:
- Type I: 0.055 µS/cm compensated to 25 °C
Type III: <20 µS/cm (depending on feed water) - Type I: 0.055 µS/cm compensated to 25 °C
Type III: <20 µS/cm (depending on feed water)
- TOC content:
- <10 ppb (system without UV lamp)
- <5 ppb (system with UV lamp)
- Resistivity at +25 °C:
- Type I: 18.2 MΩxcm & < 20 µS/cm
Type III: <0.05 MΩxcm (depending on feed water) - Type I: 18.2 MΩxcm & < 20 µS/cm
Type III: <0.05 MΩxcm (depending on feed water)
- Particles:
- <1/ml
- <1/ml
- Microorganisms:
- <1 CFU/1000 ml
- <1 CFU/1,000 ml
- W×D×H:
- 280×530.7×509.4 mm
- 280×530.7×509.4 mm
- Operating weight with full tank:
- 23 kg
- 23 kg
- Weight:
- 13 kg
- 13 kg
Frequently Bought Together