Wire Net Basket Lid, Sklar®
Fournisseur: Sklar
Certificats
A propos de cet article
All Sklar products are industry standard.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
Sklar's® Wire Net Basket Lid is used with wire net baskets of equal size. They are designed for use with cleaning, sterilizing, and handling small instruments.
Ordering information: Lids and baskets are sold separately.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.