OIML Class F1 Precision Weights, Mettler Toledo
Supplier: Mettler Toledo
Certificates
About this item
Basic Line weights are protected in robust and easy to clean plastic boxes.
- Made of stainless steel
- Adjustable cavity (AC) design
- Economical
FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.
Customers Who Bought This Also Bought