Funnels with Standard Stems, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
These clear funnels have long stems, smooth walls, and external ribs to prevent air lock.
- Autoclavable
- Molded in polypropylene
These long stemmed funnels allow you to fill tall bottles.
Customers Who Bought This Also Bought