Carboys with Stopcocks, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
- Autoclavable
- Preassembled leak-proof stopcock
These polypropylene carboys are ideal for storing and dispensing various laboratory solutions. Threads should be totally disengaged before autoclaving. The mouth Int.Ø is 67 mm.
Customers Who Bought This Also Bought