Nunc™ High Density Cell Factory™ Systems, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
High density cell factory systems provide a simple solution for increasing cell culture yields.
- 30% more surface area and yield within the standard system footprint
- Increase labor and handling efficiencies by achieving more output in a single process run
- Increase manufacturing capacity
Its has additional layers. Cell factory systems are designed to optimize any process by increasing output, while maintaining the same manufacturing footprint as with the standard cell factory system.
Consume less and reduce decontamination and waste disposal costs.
Customers Who Bought This Also Bought