Swingsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Fournisseur: Simport Scientific
Certificats
A propos de cet article
Secure specimens during processing with the Swingsette™ Biopsy Processing/Embedding Cassette, in Quickload™ stacks.
- In Quickload™ stacks
- 45° anterior writing area
- Easy to assemble hinged lids
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.