A propos de cet article
The select-a-size DNA MagBead kit provides the fastest and easiest method to purify specific ranges of DNA fragments from PCR, endonuclease digestions, ligations, library preparations, adapter removal, etc.
- Size selection can be tuned from 100 bp to 1000 bp
- Elution volume: ≥10 µl of DNA Elution Buffer
- Eluted DNA is well suited for use in next generation sequencing, PCR, DNA ligation, endonuclease digestion, etc
This allows for specific cutoffs that can be modified to suit reaction clean-up, left-sided, right-sided or even double-sided size selection. The Select-a-Size MagBeads selectively binds fragments based on the volume of buffer added relative to the sample. Simply choose a desired cutoff to bind the target of interest onto the beads and remove species outside of this range. The desired DNA is easily eluted from the beads following a rapid wash regimen.
Size selections can be performed in as little as ten minutes to yield high-quality DNA which is suitable for highly sensitive applications including Next Generation Sequencing, and any other sensitive downstream applications. The protocol can be performed manually or using an automated platform for high throughput processing.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 76197-066
- 76197-140
- Particle size:
- ≥100 bp
- ≥100 bp