Separatory Funnel Holder, United Scientific Supplies
# de catalogue 76263-260
Fournisseur: United Scientific Supplies
Spécifications
- Description:Separatory Funnel Holder
- Cat. no.:76263-260
Spécifications
A propos de cet article
Open design allows an unobstructed view.
- Tapered wedges insure a firm grip
- Front opening for an unobstructed view
This non-corrosive separatory funnel holder, molded in polypropylene, has a front opening which provides clear view of the solution and allows unobstructed placement of separating funnels. Two tapered wedges are provided to ensure firm grip on rods measuring 1/2 or 3/8" diameter.
Customers Who Bought This Also Bought