ContactUsLinkComponent
Double Funnel Holder, United Scientific Supplies
Double Funnel Holder, United Scientific Supplies
# de catalogue 76263-252
Funnel Holder 1/2 or 3/8 in
Double Funnel Holder, United Scientific Supplies
# de catalogue 76263-252
Spécifications
  • Description:
    Double Funnel Holder
  • Cat. no.:
    76263-252
Spécifications
A propos de cet article
Suspend two funnels simultaneously using this polypropylene holder.
  • Corrosion-resistant
These holders can be used with 3 to 6" diameter funnels, and include an extra plate to accommodate 1 to 3" diameter funnels. Two tapered wedges are provided to ensure firm grip on rods measuring 1/2 or 3/8" diameter.
Customers Who Bought This Also Bought