TYE Agar Plates, with Glucose, Teknova
Fournisseur: Teknova
Certificats
A propos de cet article
It contains 1.0% Tryptone, 0.8% Sodium chloride, 0.5% Yeast extract, 0.1 or 1.0% Glucose, 1.5% agar.
- Store at room temperature
- Sterile
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.