A propos de cet article
Clean and sterile disposable coveralls are individually double-bagged and folded to promote easier gowning with less excursions.
- Clean and sterile
- Manufactured in silicone-free environment
- Latex-free
- Tunneled elastic on wrists (with thumb loops), ankles, and back half of waist
- Individually double-bagged
- Certificate of radiation included
- Gamma radiation indicator dots on each package
CleanMax® cleanroom garments provide comfort, quality, and protection. CleanMax® sterilized garments are both clean manufactured and sterile (not all sterile garments are clean manufactured).
All these apparels provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunneled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Spécifications
- Catalog No:
- 76193-328
- 76193-330
- 76193-334
- 76193-336
- 76193-338
- 76193-340
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom