VWR® Cap Mat
Fournisseur: VWR International
Certificats
A propos de cet article
Re-usable cap mats are available for friction sealing of plates.
Cap mats are an economic alternative to thermal or adhesive seals as they can be sterilized and reused.
Customers Who Bought This Also Bought