DURAN® YOUTILITY® GL 45 Laboratory Bottle System, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
The new generation of glass laboratory bottles.
- Borosilicate 3.3 pharmacopeia type 1 neutral glass bottles
- Bottle tags are made of silicone and available in eight colors
- Labels accept copier, printer, ballpoint, and technical markers and inks
- Pour rings are autoclavable hydrophilic polypropylene
The updated design includes an improved ergonomic bottle shape and dedicated accessories for better handling, sample identification, and ease of use. The GL 45 bottles have a defined labelling area and an integrated retrace code for full traceability.
Customers Who Bought This Also Bought