KimCote® Solution Bottles with Color-Coded PTFE Flathead Stoppers, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
KimCote® solution bottles are designed for storage of solutions and ideal for media preparations.
- Narrow mouth increases mechanical strength
- Bottle necks are standard taper ground to accept flathead color-coded PTFE stoppers
- Manufactured from 33 expansion, low extractable borosilicate glass
- Replacement stoppers are 41941
Autoclavable safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals.
Certifications: Meets USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought