ContactUsLinkComponent
KIMBLE® Lab Starter Kit, DWK Life Sciences
KIMBLE® Lab Starter Kit, DWK Life Sciences
# de catalogue 76176-856
KIMBLE® Lab Starter Kit, DWK Life Sciences
# de catalogue 76176-856
Spécifications
  • Description:
    Lab Starter Kit
  • Cat. no.:
    76176-856
Spécifications
A propos de cet article
The lab starter kit includes all the glassware needed to get started.
Ordering information: Kit includes low form griffin beaker (50 mL×3, 100 mL×3, 250 mL×6), wide mouth erlenmeyer flask (250 mL×3, 500 mL×6, 1000 mL×3), class B cylinders with single metric scale (100 mL, 250 mL, 500 mL each), GL 45 media bottle (100 mL×2, 250×3, 500 mL×3, 1000 mL×2).
Customers Who Bought This Also Bought